Assay Details
Gene Symbol: LOC114675648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842841.1 | XP_028698674.1 | 157 | 86 |
Gene Symbol: GABRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261778.1 | NP_001248707.1 | 1191 | 86 | ||
XM_015141355.2 | XP_014996841.1 | 1601 | 86 | |||
GenBank mRNA | JU320012.1 | - | 1191 | 86 | ||
JU320013.1 | - | 1191 | 86 | |||
JV489518.1 | - | 1191 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
