Assay Details
| Gene Symbol: MNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266861.1 | NP_001253790.1 | 955 | 67 | ||
| GenBank mRNA | JU473226.1 | - | 962 | 67 | ||
| JV047559.1 | - | 955 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MNS1
Genomic Map