Assay Details
Gene Symbol: KASH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124565.2 | XP_014980051.1 | 1208 | 60 | ||
XM_015124566.2 | XP_014980052.1 | 1208 | 60 | |||
XM_015124568.2 | XP_014980054.1 | 1431 | 60 | |||
XM_015124570.2 | XP_014980056.1 | 1263 | 60 | |||
XM_028838960.1 | XP_028694793.1 | 2398 | 60 | |||
XM_028838961.1 | XP_028694794.1 | 2230 | 60 | |||
XM_028838962.1 | XP_028694795.1 | 2326 | 60 | |||
XM_028838963.1 | XP_028694796.1 | 2398 | 60 | |||
XM_028838965.1 | XP_028694798.1 | 1376 | 60 | |||
GenBank mRNA | JV052633.1 | - | 340 | 60 | ||
JV643157.1 | - | 392 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
