Assay Details
| Gene Symbol: MRPS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257635.1 | NP_001244564.1 | 283 | 71 | ||
| XM_015136374.2 | XP_014991860.1 | 326 | 71 | |||
| XM_028846714.1 | XP_028702547.1 | 324 | 71 | |||
| GenBank mRNA | JU335248.1 | - | 283 | 71 | ||
| JU476220.1 | - | 281 | 71 | |||
| JV047514.1 | - | 270 | 71 | |||
| JV047515.1 | - | 270 | 71 | |||
| JV549595.1 | - | 283 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPS18A
Genomic Map