Assay Details
Gene Symbol: LOC715257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110742.2 | XP_014966228.2 | 1044 | 150 | ||
XM_015110743.2 | XP_014966229.2 | 1084 | 150 | |||
XM_015110744.2 | XP_014966230.2 | 907 | 150 | |||
XM_015110745.2 | XP_014966231.2 | 786 | 150 | |||
XM_028831002.1 | XP_028686835.1 | 1327 | 150 | |||
XM_028831003.1 | XP_028686836.1 | 932 | 150 | |||
GenBank mRNA | JU326862.1 | - | 692 | 150 | ||
JU476529.1 | - | 817 | 150 | |||
JV635566.1 | - | 1351 | 150 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
