Assay Details
| Gene Symbol: PPP2R1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257922.1 | NP_001244851.1 | 240 | 69 | ||
| GenBank mRNA | JU319962.1 | - | 240 | 69 | ||
| JU470550.1 | - | 352 | 69 | |||
| JV047520.1 | - | 257 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP2R1A
Genomic Map