Assay Details
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193883.2 | NP_001180812.2 | 723 | 81 | ||
XM_015110595.2 | XP_014966081.2 | 961 | 81 | |||
XM_015110596.2 | XP_014966082.2 | 961 | 81 | |||
XM_015110597.2 | XP_014966083.1 | 872 | 81 | |||
XM_028830441.1 | XP_028686274.1 | 924 | 81 | |||
GenBank mRNA | JU338818.1 | - | 703 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
