Assay Details
| Gene Symbol: DAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266812.1 | NP_001253741.1 | 159 | 80 | ||
| XM_015122454.2 | XP_014977940.1 | 297 | 80 | |||
| XM_015122455.2 | XP_014977941.1 | 254 | 80 | |||
| GenBank mRNA | JU333177.1 | - | 153 | 80 | ||
| JU472653.1 | - | 159 | 80 | |||
| JV046304.1 | - | 159 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DAPK3
Genomic Map