Assay Details
| Gene Symbol: PPP1CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260652.2 | NP_001247581.1 | 374 | 62 | ||
| XM_015111926.2 | XP_014967412.1 | 765 | 62 | |||
| GenBank mRNA | JU335184.1 | - | 202 | 62 | ||
| JU335185.1 | - | 202 | 62 | |||
| JU335186.1 | - | 202 | 62 | |||
| JU335187.1 | - | 202 | 62 | |||
| JU471793.1 | - | 200 | 62 | |||
| JU471794.1 | - | 256 | 62 | |||
| JU471795.1 | - | 256 | 62 | |||
| JV043567.1 | - | 370 | 62 | |||
| JV047480.1 | - | 374 | 62 | |||
| JV047481.1 | - | 374 | 62 | |||
| JV654851.1 | - | 213 | 62 | |||
More Information
Set Membership: |
|

PPP1CB
Genomic Map