Assay Details
Gene Symbol: TRAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118931.1 | XP_014974417.1 | 314 | 77 | ||
XM_028836153.1 | XP_028691986.1 | 131 | 77 | |||
GenBank mRNA | JU321792.1 | - | 282 | 77 | ||
JU473096.1 | - | 279 | 77 | |||
JV043916.1 | - | 248 | 77 | |||
JV636553.1 | - | 296 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028836153.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
