Assay Details
Gene Symbol: ARPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261239.1 | NP_001248168.1 | 555 | 60 | ||
XM_015110982.2 | XP_014966468.1 | 704 | 60 | |||
GenBank mRNA | JU320784.1 | - | 624 | 60 | ||
JU332406.1 | - | 624 | 60 | |||
JU473869.1 | - | 625 | 60 | |||
JU473870.1 | - | 625 | 60 | |||
JU473871.1 | - | 625 | 60 | |||
JU473872.1 | - | 625 | 60 | |||
JV044180.1 | - | 555 | 60 | |||
JV602938.1 | - | 792 | 60 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
