Assay Details
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139315.2 | XP_014994801.1 | 2361 | 96 | ||
XM_028848798.1 | XP_028704631.1 | 2373 | 96 | |||
XM_028848799.1 | XP_028704632.1 | 2258 | 96 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
