Assay Details
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194341.2 | NP_001181270.1 | 308 | 92 | ||
XM_015116531.2 | XP_014972017.1 | 248 | 92 | |||
GenBank mRNA | JV704188.1 | - | 287 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
