Assay Details
Gene Symbol: VPS26C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265971.1 | NP_001252900.1 | 643 | 149 | ||
GenBank mRNA | JU322030.1 | - | 750 | 149 | ||
JU332351.1 | - | 750 | 149 | |||
JU332352.1 | - | 750 | 149 | |||
JU473631.1 | - | 643 | 149 | |||
JV045882.1 | - | 716 | 149 | |||
JV045883.1 | - | 716 | 149 | |||
JV045885.1 | - | 716 | 149 | |||
JV045886.1 | - | 716 | 149 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
