Assay Details
Gene Symbol: NPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266786.1 | NP_001253715.1 | 885 | 61 | ||
XM_015142608.2 | XP_014998094.1 | 920 | 61 | |||
XM_015142609.1 | XP_014998095.1 | 559 | 61 | |||
XM_015142610.1 | XP_014998096.1 | 559 | 61 | |||
XM_028850261.1 | XP_028706094.1 | 675 | 61 | |||
GenBank mRNA | JU327952.1 | - | 644 | 61 | ||
JU327953.1 | - | 644 | 61 | |||
JU473037.1 | - | 547 | 61 | |||
JV045602.1 | - | 462 | 61 | |||
JV613190.1 | - | 906 | 61 | |||
JV636098.1 | - | 430 | 61 | |||
JV667373.1 | - | 885 | 61 | |||
JV667374.1 | - | 885 | 61 | |||
JV667375.1 | - | 537 | 61 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
