Assay Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120495.2 | XP_014975981.2 | 311 | 62 | ||
XM_028836879.1 | XP_028692712.1 | 211 | 62 | |||
GenBank mRNA | JU477531.1 | - | 245 | 62 | ||
JV048475.1 | - | 209 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028836879.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
