Assay Details
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133230.2 | XP_014988716.1 | 1921 | 70 | ||
GenBank mRNA | JU336473.1 | - | 1799 | 70 | ||
JU477283.1 | - | 1800 | 70 | |||
JV521035.1 | - | 201 | 70 | |||
JV600819.1 | - | 1830 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
