Assay Details
| Gene Symbol: BCAP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128516.2 | XP_014984002.1 | 782 | 65 | ||
| XM_028842309.1 | XP_028698142.1 | 446 | 65 | |||
| XM_028842310.1 | XP_028698143.1 | 681 | 65 | |||
| GenBank mRNA | JU326533.1 | - | 212 | 65 | ||
| JU478528.1 | - | 392 | 65 | |||
| JV530363.1 | - | 388 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCAP31
Genomic Map