Assay Details
Gene Symbol: SRPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190876.1 | NP_001177805.1 | 152 | 66 | ||
XM_015132068.2 | XP_014987554.1 | 529 | 66 | |||
XM_028843441.1 | XP_028699274.1 | 584 | 66 | |||
XM_028843442.1 | XP_028699275.1 | 431 | 66 | |||
GenBank mRNA | JU335721.1 | - | 207 | 66 | ||
JU476609.1 | - | 232 | 66 | |||
JV589438.1 | - | 165 | 66 | |||
JV691972.1 | - | 166 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
