Assay Details
Gene Symbol: SLC16A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260620.1 | NP_001247549.1 | 429 | 63 | ||
GenBank mRNA | JU471273.1 | - | 429 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
