Assay Details
Gene Symbol: SOX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099185.3 | XP_001099185.1 | 1389 | 94 | ||
XM_015119752.2 | XP_014975238.1 | 903 | 94 | |||
XM_015119755.2 | XP_014975241.1 | 888 | 94 | |||
GenBank mRNA | JU337230.1 | - | 894 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
