Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260939.1 | NP_001247868.1 | 517 | 114 | ||
| GenBank mRNA | JU335502.1 | - | 517 | 114 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF5A2| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260939.1 | NP_001247868.1 | 517 | 114 | ||
| GenBank mRNA | JU335502.1 | - | 517 | 114 | ||
Set Membership: |
|