Assay Details
| Gene Symbol: AATF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257577.1 | NP_001244506.1 | 1649 | 115 | ||
| GenBank mRNA | JU333969.1 | - | 1649 | 115 | ||
| JU475152.1 | - | 1633 | 115 | |||
| JV046741.1 | - | 1661 | 115 | |||
| JV680300.1 | - | 970 | 115 | |||
More Information
Set Membership: |
|

AATF
Genomic Map