Assay Details
| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266189.1 | NP_001253118.1 | 1224 | 79 | ||
| XM_015135211.2 | XP_014990697.1 | 1234 | 79 | |||
| XM_028845059.1 | XP_028700892.1 | 1205 | 79 | |||
| XM_028845060.1 | XP_028700893.1 | 1425 | 79 | |||
| XM_028845061.1 | XP_028700894.1 | 1277 | 79 | |||
| GenBank mRNA | JU319934.1 | - | 1224 | 79 | ||
| JU472393.1 | - | 1244 | 79 | |||
| JV690597.1 | - | 1262 | 79 | |||
More Information
Set Membership: |
|

ACTR3B
Genomic Map