Assay Details
| Gene Symbol: UBE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194143.2 | NP_001181072.1 | 760 | 80 | ||
| GenBank mRNA | JU472964.1 | - | 139 | 80 | ||
| JV632821.1 | - | 142 | 80 | |||
More Information
Set Membership: |
|

UBE2M
Genomic Map