Assay Details
| Gene Symbol: NTNG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265764.1 | NP_001252693.1 | 1956 | 85 | ||
| XM_015116494.2 | XP_014971980.2 | 2155 | 85 | |||
| XM_028834382.1 | XP_028690215.1 | 2411 | 85 | |||
| XM_028834383.1 | XP_028690216.1 | 2435 | 85 | |||
| XM_028834384.1 | XP_028690217.1 | 2333 | 85 | |||
| XM_028834385.1 | XP_028690218.1 | 2363 | 85 | |||
| XM_028834386.1 | XP_028690219.1 | 2247 | 85 | |||
| XM_028834387.1 | XP_028690220.1 | 2213 | 85 | |||
| XM_028834388.1 | XP_028690221.1 | 2496 | 85 | |||
| GenBank mRNA | JU336755.1 | - | 1928 | 85 | ||
| JV044847.1 | - | 1973 | 85 | |||
| JV667287.1 | - | 1956 | 85 | |||
More Information
Set Membership: |
|

NTNG2
Genomic Map