Assay Details
| Gene Symbol: MAN1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137511.2 | XP_014992997.1 | 1195 | 99 | ||
| XM_015137512.2 | XP_014992998.1 | 1264 | 99 | |||
| GenBank mRNA | JU326917.1 | - | 240 | 99 | ||
| JV565120.1 | - | 1244 | 99 | |||
| JV715088.1 | - | 740 | 99 | |||
More Information
Set Membership: |
|

MAN1A1
Genomic Map