Assay Details
Gene Symbol: SEPTIN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138470.2 | XP_014993956.1 | 843 | 63 | ||
XM_015138471.2 | XP_014993957.1 | 843 | 63 | |||
XM_015138472.2 | XP_014993958.1 | 843 | 63 | |||
XM_015138473.2 | XP_014993959.1 | 907 | 63 | |||
XM_015138474.2 | XP_014993960.1 | 843 | 63 | |||
XM_015138476.2 | XP_014993962.1 | 843 | 63 | |||
XM_015138477.2 | XP_014993963.1 | 843 | 63 | |||
XM_028848482.1 | XP_028704315.1 | 859 | 63 | |||
GenBank mRNA | JU335418.1 | - | 826 | 63 | ||
JU335419.1 | - | 826 | 63 | |||
JU472070.1 | - | 856 | 63 | |||
JU472071.1 | - | 856 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
