Assay Details
Gene Symbol: LMNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260848.1 | NP_001247777.1 | 1379 | 79 | ||
GenBank mRNA | JU327450.1 | - | 501 | 79 | ||
JU471250.1 | - | 1379 | 79 | |||
JV635481.1 | - | 1489 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
