Assay Details
| Gene Symbol: ASIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271252.1 | NP_001258181.1 | 1599 | 59 | ||
| XM_028835771.1 | XP_028691604.1 | 2461 | 59 | |||
| GenBank mRNA | JU327464.1 | - | 1058 | 59 | ||
| JU327515.1 | - | 1270 | 59 | |||
| JV468449.1 | - | 1288 | 59 | |||
| JV544783.1 | - | 1078 | 59 | |||
| JV581692.1 | - | 1362 | 59 | |||
| JV581693.1 | - | 1386 | 59 | |||
| JV683896.1 | - | 1121 | 59 | |||
| JV714874.1 | - | 975 | 59 | |||
More Information
Set Membership: |
|

ASIC2
Genomic Map