Assay Details
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265906.1 | NP_001252835.1 | 334 | 69 | ||
XM_028834279.1 | XP_028690112.1 | 286 | 69 | |||
GenBank mRNA | JU319899.1 | - | 314 | 69 | ||
JU473563.1 | - | 337 | 69 | |||
JV047532.1 | - | 334 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
