Assay Details
| Gene Symbol: CHRNA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032883.1 | NP_001028055.1 | 342 | 101 | ||
| GenBank mRNA | AF486623.1 | - | 342 | 101 | ||
| JV604943.1 | - | 314 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CHRNA7
Genomic Map