Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265687.1 | NP_001252616.1 | 7390 | 88 | ||
| XM_015140180.2 | XP_014995666.2 | 7376 | 88 | |||
| GenBank mRNA | AY369829.1 | - | 705 | 88 | ||
| JU326263.1 | - | 5161 | 88 | |||
| JU326264.1 | - | 5161 | 88 | |||
| JU326265.1 | - | 5161 | 88 | |||
| JU326266.1 | - | 5161 | 88 | |||
| JU326267.1 | - | 5161 | 88 | |||
| JU326268.1 | - | 5161 | 88 | |||
| JV564958.1 | - | 7390 | 88 | |||
| JV616851.1 | - | 7461 | 88 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAP1B
Genomic Map