Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257671.2 | NP_001244600.1 | 670 | 89 | ||
| NM_001320538.1 | NP_001307467.1 | 670 | 89 | |||
| GenBank mRNA | JU334370.1 | - | 669 | 89 | ||
| JU334371.1 | - | 669 | 89 | |||
| JV451890.1 | - | 571 | 89 | |||
| JV451891.1 | - | 571 | 89 | |||
| JV505564.1 | - | 590 | 89 | |||
| JV528281.1 | - | 668 | 89 | |||
| JV580751.1 | - | 668 | 89 | |||
| JV682891.1 | - | 668 | 89 | |||
| JV714097.1 | - | 670 | 89 | |||
More Information
Set Membership: |
|

AQP4
Genomic Map