Assay Details
Gene Symbol: BVES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266212.1 | NP_001253141.1 | 984 | 105 | ||
XM_028846641.1 | XP_028702474.1 | 1140 | 105 | |||
GenBank mRNA | JU321484.1 | - | 984 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
