Assay Details
Gene Symbol: CDKN2AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261581.1 | NP_001248510.1 | 431 | 81 | ||
XM_015139572.2 | XP_014995058.2 | 418 | 81 | |||
XM_015139573.2 | XP_014995059.1 | 697 | 81 | |||
GenBank mRNA | JU322461.1 | - | 425 | 81 | ||
JU470704.1 | - | 431 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015139572.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
