Assay Details
| Gene Symbol: AP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261143.2 | NP_001248072.1 | 369 | 70 | ||
| GenBank mRNA | JU332852.1 | - | 304 | 70 | ||
| JU474251.1 | - | 369 | 70 | |||
| JV532864.1 | - | 364 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP2S1
Genomic Map