Assay Details
| Gene Symbol: BNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111612.4 | XP_001111612.2 | 420 | 80 | ||
| XM_015142893.2 | XP_014998379.2 | 285 | 80 | |||
| GenBank mRNA | JV658887.1 | - | 214 | 80 | ||
More Information
Set Membership: |
|

BNC1
Genomic Map