Assay Details
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836815.1 | XP_028692648.1 | 1012 | 67 | ||
XM_028836816.1 | XP_028692649.1 | 1164 | 67 | |||
XM_028836817.1 | XP_028692650.1 | 890 | 67 | |||
XM_028836818.1 | XP_028692651.1 | 795 | 67 | |||
XM_028836819.1 | XP_028692652.1 | 1012 | 67 | |||
XM_028836820.1 | XP_028692653.1 | 1012 | 67 | |||
XM_028836821.1 | XP_028692654.1 | 1012 | 67 | |||
XM_028836822.1 | XP_028692655.1 | 1116 | 67 | |||
XM_028836823.1 | XP_028692656.1 | 1072 | 67 | |||
XM_028836824.1 | XP_028692657.1 | 1002 | 67 | |||
XM_028836825.1 | XP_028692658.1 | 815 | 67 | |||
XM_028836826.1 | XP_028692659.1 | 477 | 67 | |||
GenBank mRNA | JV637311.1 | - | 127 | 67 | ||
JV704971.1 | - | 89 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
