Assay Details
| Gene Symbol: GPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118462.2 | XP_014973948.1 | 619 | 101 | ||
| XM_028835957.1 | XP_028691790.1 | 516 | 101 | |||
| XM_028835958.1 | XP_028691791.1 | 522 | 101 | |||
| GenBank mRNA | JU327335.1 | - | 179 | 101 | ||
| JU473330.1 | - | 399 | 101 | |||
| JV528798.1 | - | 383 | 101 | |||
| JV635654.1 | - | 395 | 101 | |||
More Information
Set Membership: |
|

GPS2
Genomic Map