Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101117.4 | XP_001101117.1 | 173 | 72 | ||
| GenBank mRNA | JV600163.1 | - | 69 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001101117.4 transcripts.
Set Membership: |
|

TUBA4A
Genomic Map