Assay Details
| Gene Symbol: L2HGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099959.4 | XP_001099959.1 | 646 | 128 | ||
| XM_015143658.2 | XP_014999144.1 | 710 | 128 | |||
| XM_028851537.1 | XP_028707370.1 | 646 | 128 | |||
More Information
Set Membership: |
|

L2HGDH
Genomic Map