Assay Details
| Gene Symbol: HNRNPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260807.2 | NP_001247736.1 | 930 | 105 | ||
| XM_015138420.2 | XP_014993906.1 | 899 | 105 | |||
| XM_015138421.2 | XP_014993907.1 | 935 | 105 | |||
| XM_015138422.2 | XP_014993908.1 | 886 | 105 | |||
| XM_028847966.1 | XP_028703799.1 | 807 | 105 | |||
| XM_028847967.1 | XP_028703800.1 | 923 | 105 | |||
| GenBank mRNA | JU319927.1 | - | 878 | 105 | ||
| JU319928.1 | - | 935 | 105 | |||
| JU319929.1 | - | 935 | 105 | |||
| JU475390.1 | - | 589 | 105 | |||
| JU475391.1 | - | 646 | 105 | |||
| JU475392.1 | - | 646 | 105 | |||
| JV043558.1 | - | 878 | 105 | |||
| JV043559.1 | - | 935 | 105 | |||
| JV043560.1 | - | 935 | 105 | |||
| JV637643.1 | - | 885 | 105 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNRNPD
Genomic Map