Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257695.2 | NP_001244624.1 | 1326 | 78 | ||
| XM_028841906.1 | XP_028697739.1 | 1458 | 78 | |||
| GenBank mRNA | JU332740.1 | - | 1326 | 78 | ||
More Information
Set Membership: |
|

DUSP10
Genomic Map