Assay Details
Gene Symbol: SAP30BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261024.1 | NP_001247953.1 | 635 | 72 | ||
XM_015120325.2 | XP_014975811.1 | 779 | 72 | |||
XM_015120326.2 | XP_014975812.1 | 961 | 72 | |||
XM_015120327.2 | XP_014975813.1 | 741 | 72 | |||
XM_015120328.2 | XP_014975814.1 | 628 | 72 | |||
XM_015120329.2 | XP_014975815.1 | 811 | 72 | |||
XM_028835535.1 | XP_028691368.1 | 778 | 72 | |||
XM_028835536.1 | XP_028691369.1 | 1080 | 72 | |||
XM_028835537.1 | XP_028691370.1 | 1000 | 72 | |||
GenBank mRNA | JU321642.1 | - | 632 | 72 | ||
JU475293.1 | - | 635 | 72 | |||
JV579611.1 | - | 645 | 72 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
