Assay Details
| Gene Symbol: TMEM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120447.2 | XP_014975933.1 | 420 | 74 | ||
| XM_015120453.2 | XP_014975939.1 | 423 | 74 | |||
| XM_015120461.2 | XP_014975947.1 | 708 | 74 | |||
| XM_015120469.2 | XP_014975955.1 | 404 | 74 | |||
| XM_028851981.1 | XP_028707814.1 | 540 | 74 | |||
| XM_028851984.1 | XP_028707817.1 | 512 | 74 | |||
| GenBank mRNA | JU333770.1 | - | 312 | 74 | ||
| JU333771.1 | - | 303 | 74 | |||
| JU472997.1 | - | 277 | 74 | |||
| JV046638.1 | - | 353 | 74 | |||
| JV637534.1 | - | 284 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM9
Genomic Map