Assay Details
Gene Symbol: DES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194266.2 | NP_001181195.1 | 1500 | 78 | ||
GenBank mRNA | JU327795.1 | - | 792 | 78 | ||
JU481567.1 | - | 1373 | 78 | |||
JV644043.1 | - | 1339 | 78 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
