Assay Details
| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265937.2 | NP_001252866.1 | 547 | 78 | ||
| XM_015143344.2 | XP_014998830.1 | 665 | 78 | |||
| XM_028850494.1 | XP_028706327.1 | 627 | 78 | |||
| GenBank mRNA | JV047233.1 | - | 545 | 78 | ||
| JV635486.1 | - | 547 | 78 | |||
More Information
Set Membership: |
|

PCK2
Genomic Map