Assay Details
| Gene Symbol: PACSIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266225.1 | NP_001253154.1 | 823 | 56 | ||
| XM_028832714.1 | XP_028688547.1 | 944 | 56 | |||
| XM_028832715.1 | XP_028688548.1 | 811 | 56 | |||
| GenBank mRNA | JU320821.1 | - | 823 | 56 | ||
More Information
Set Membership: |
|

PACSIN3
Genomic Map