Assay Details
Gene Symbol: CACNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124803.2 | XP_014980289.2 | 550 | 54 | ||
GenBank mRNA | JV692211.1 | - | 387 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
